1-Cyanomethyl-2-(3-cyanophenyl)-4,5-dicyanoimidazole ≥97%

Supplier: Thermo Scientific

H54024.06EA 594 EUR
H54024.06 H54024.MD H54024.03
1-Cyanomethyl-2-(3-cyanophenyl)-4,5-dicyanoimidazole ≥97%
1-Cyanomethyl-2-(3-cyanophenyl)-4,5-dicyanoimidazole
Formula: C₁₄H₆N₆
Storage Temperature: Ambient
MDL Number: MFCD20265160
CAS Number: 1347815-42-2
UN: 3439
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR