4-Bromo-3-methyl-1H-pyrazole 97%

Supplier: Thermo Scientific

Warning

Synonyms: 4-Bromo-3-methylpyrazole

ACRO344350010EA 49.4 EUR
ACRO344350010
4-Bromo-3-methyl-1H-pyrazole 97%
4-Bromo-3-methyl-1H-pyrazole
Formula: C₄H₅BrN₂
MW: 161,01
Storage Temperature: Ambient
MDL Number: MFCD00005241
CAS Number: 13808-64-5
EINECS: 237-456-7

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Melting point 72 to 79 °C
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR