3-Amino-4-bromo-1-methyl-1H-pyrazole ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-Bromo-1-methyl-1H-pyrazol-3-amine
B21592.03EA
111
EUR
B21592.03
B21592.06
3-Amino-4-bromo-1-methyl-1H-pyrazole ≥97%
3-Amino-4-bromo-1-methyl-1H-pyrazole
Formula:
C₄H₆BrN₃ MW: 176,02 g/mol Melting Pt: 96…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00663065 CAS Number: 146941-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...