4-Bromo-1,3,5-trimethyl-1H-pyrazole 95%

Supplier: Thermo Scientific

Synonyms: 4-Bromo-1,3,5-trimethylpyrazole

L11876.06EA 133 EUR
L11876.06 L11876.03
4-Bromo-1,3,5-trimethyl-1H-pyrazole 95%
4-Bromo-1,3,5-trimethyl-1H-pyrazole
Formula: C₆H₉BrN₂
MW: 189,06
Boiling Pt: 101…102 °C (15 mmHg)
Melting Pt: 32…37 °C
Storage Temperature: Ambient
MDL Number: MFCD00052532
CAS Number: 15801-69-1
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR