1,3,5-Trimethyl-1H-pyrazole-4-sulphonylchloride

MAYBCC14703CBEA 0 EUR
MAYBCC14703CB MAYBCC14703DA
1,3,5-Trimethyl-1H-pyrazole-4-sulphonylchloride
1,3,5-Trimethyl-1H-pyrazole-4-sulphonylchloride
Formula: C₆H₉ClN₂O₂S
MDL Number: MFCD02180944
CAS Number: 59340-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR