4'-(Phenylethynyl)acetophenone ≥97%

Supplier: Thermo Scientific

A19370.06EA 0 EUR
A19370.06
4'-(Phenylethynyl)acetophenone ≥97%
4'-(Phenylethynyl)acetophenone
Formula: C₁₆H₁₂O
Melting Pt: 99…102 °C
MDL Number: MFCD00219774
CAS Number: 1942-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR