Tris[N,N-bis(trimethylsilyl)amide]erbium(III) 98%

Supplier: Acros Organics
Danger

ACRO381580010EA 108 EUR
ACRO381580010
Tris[N,N-bis(trimethylsilyl)amide]erbium(III) 98%
Tris[N,N-bis(trimethylsilyl)amide]erbium (III)
Formula: C₁₈H₅₄ErN₃Si₆
Melting Pt: 180…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00271002
CAS Number: 103457-72-3
UN: 3396
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Pink Crystals
Elemental Analysis 32.5 to 34 % (C)
Elemental Analysis 8.2 to 8.6 % (H)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR