4-(1,3-Oxazol-5-yl)aniline
MAYBDFP00120CBEA
0
EUR
MAYBDFP00120.0100
MAYBDFP00120CB
MAYBDFP00120DA
4-(1,3-Oxazol-5-yl)aniline
4-(1,3-Oxazol-5-yl)aniline
Formula:
C₉H₈N₂O MW: 160,1700 g/mol |
MDL Number:
MFCD00052192 CAS Number: 1008-95-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...