(±)-Naringenin
Supplier: Molekula
Synonyms:
(±)-2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one,5,7-Dihydroxy-2-(4-hydroxyphenyl)chroman-4-one,4',5,7-Trihydroxyflavanone,Naringenin
36117021-5G
MOLE36117021-5GEA
227
EUR
MOLE36117021-5G
(±)-Naringenin
(±)-Naringenin
Formula:
C₁₅H₁₂O₅ MW: 272,25 g/mol |
MDL Number:
MFCD00006844 CAS Number: 480-41-1 EINECS: 207-550-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...