(±)-Taxifolin

Supplier: Enzo Life Sciences

ALEXALX385006M050EA 221 EUR
ALEXALX385006M050
(±)-Taxifolin
(±)-Taxifolin
Formula: C₁₅H₁₂O₇
CAS Number: 24198-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR