3-Diethylsulfamoyl-4-methoxybenzeneboronic acid, 95%

H52770.MDEA 48.6 EUR
ALFAH52770.MD ALFAH52770.03
3-Diethylsulfamoyl-4-methoxybenzeneboronic acid, 95%
Purity: 95%
CAS No.: 871333-03-8
Molecular Formula: C11H18BNO5S
Formula Weight: 287.14
Melting Point: 110-112°
MDL No.: MFCD08056367
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR