3-(Diethylsulfamoyl)benzeneboronic acid, 98%

H52593.03EA 270.5 EUR
ALFAH52593.03 ALFAH52593.MD
3-(Diethylsulfamoyl)benzeneboronic acid, 98%
Purity: 98%
CAS No.: 871329-58-7
Molecular Formula: C10H16BNO4S
Formula Weight: 257.11
Melting Point: 120-123°
MDL No.: MFCD07363746
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR