3-(n-Butylsulfamoyl)benzeneboronic acid, 95%

H52484.MDEA 144.9 EUR
ALFAH52484.MD
3-(n-Butylsulfamoyl)benzeneboronic acid, 95%
Purity: 95%
CAS No.: 871329-78-1
Molecular Formula: C10H16BNO4S
Formula Weight: 257.11
Melting Point: 92-94°
MDL No.: MFCD07783858
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR