3-(Cyclohexylsulfamoyl)benzeneboronic acid, 97%

H52915.MDEA 56.5 EUR
ALFAH52915.MD
3-(Cyclohexylsulfamoyl)benzeneboronic acid, 97%
Purity: 97%
CAS No.: 871329-79-2
Molecular Formula: C12H18BNO4S
Formula Weight: 283.15
MDL No.: MFCD07783859
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR