4-Bromo-3-ethoxyaniline hydrochloride ≥98%

Supplier: Thermo Scientific

Danger

B24827.06EA 22.6 EUR
B24827.06 B24827.22 B24827.14
4-Bromo-3-ethoxyaniline hydrochloride ≥98%
4-Bromo-3-ethoxyaniline hydrochloride
Formula: C₈H₁₁BrClNO
Melting Pt: 186…189 °C
Storage Temperature: Ambient
MDL Number: MFCD03790891
CAS Number: 846023-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR