4-(3-Chloropropylsulfamoyl)benzeneboronic acid, 98%

H53099.MDEA 76.8 EUR
ALFAH53099.MD
4-(3-Chloropropylsulfamoyl)benzeneboronic acid, 98%
Purity: 98%
CAS No.: 874219-48-4
Molecular Formula: C9H13BClNO4S
Formula Weight: 277.53
Melting Point: 243-245°
MDL No.: MFCD08235065
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR