5-Chloro-2-nitrodiphenylamine ≥98%, rods

Supplier: Alfa Aesar

B24179.14EA 0 EUR
B24179.14
5-Chloro-2-nitrodiphenylamine ≥98%, rods
5-Chloro-2-nitrodiphenylamine
Formula: C₁₂H₉ClN₂O₂
Melting Pt: 108…111 °C
MDL Number: MFCD00007287
CAS Number: 25781-92-4
EINECS: 247-261-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR