5-Chloro-2-nitrodiphenylamine

Supplier: Molekula

MOLE30181991-25GEA 0 EUR
MOLE30181991-25G
5-Chloro-2-nitrodiphenylamine
5-Chloro-2-nitrodiphenylamine
Formula: C₁₂H₉ClN₂O₂
MDL Number: MFCD00007287
CAS Number: 25781-92-4
EINECS: 247-261-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR