4-Fluoro-3-formylbenzeneboronic acid pinacol ester, 96%

H52310.MDEA 139.7 EUR
ALFAH52310.MD ALFAH52310.03
4-Fluoro-3-formylbenzeneboronic acid pinacol ester, 96%
Purity: 96%
CAS No.: 443776-94-1
Molecular Formula: C13H16BFO3
Formula Weight: 250.07
MDL No.: MFCD11846346
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR