4-Amino-3-chloro-5-methylbenzonitrile tech. 90%, Technical Grade
Supplier: Thermo Scientific
|
Danger
|
L12541.03EA
100
EUR
L12541.03
4-Amino-3-chloro-5-methylbenzonitrile tech. 90%, Technical Grade
4-Amino-3-chloro-5-methylbenzonitrile
Formula:
C₈H₇ClN₂ MW: 166,61 g/mol Melting Pt: 128…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041476 CAS Number: 158296-69-6 EINECS: 000-000-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...