N-(3,5-Dichlorophenyl)-3-(trifluoromethyl)benzamide, 97%

H57215.MDEA 0 EUR
ALFAH57215.MD ALFAH57215.03
N-(3,5-Dichlorophenyl)-3-(trifluoromethyl)benzamide, 97%
Purity: 97%
Molecular Formula: C14H8Cl2F3NO
Formula Weight: 334.12
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR