2,3-Dichlorobenzamidoxime

MAYBSPB01106DAEA 0 EUR
MAYBSPB01106DA MAYBSPB01106CB
2,3-Dichlorobenzamidoxime
2,3-Dichlorobenzamidoxime
Formula: C₇H₆Cl₂N₂O
MDL Number: MFCD01567248
CAS Number: 261761-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR