2,6-Dichlorobenzamidoxime

MAYBSPB00002DAEA 0 EUR
MAYBSPB00002DA
2,6-Dichlorobenzamidoxime
2,6-Dichlorobenzamidoxime
Formula: C₇H₆Cl₂N₂O
MDL Number: MFCD00221076
CAS Number: 23505-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR