4-Hydroxy-3-(trifluoromethyl)benzaldehyde ≥98%

Supplier: Thermo Scientific
Warning

H26098.03EA 304 EUR
H26098.03 H26098.MD
4-Hydroxy-3-(trifluoromethyl)benzaldehyde ≥98%
4-Hydroxy-3-(trifluoromethyl)benzaldehyde
Formula: C₈H₅F₃O₂
Melting Pt: 159…163 °C
Storage Temperature: Ambient
MDL Number: MFCD01091006
CAS Number: 220227-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR