N-(3,5-Difluorophenyl)-2-(trifluoromethyl)benzamide, 97%

H57987.03EA 180.8 EUR
ALFAH57987.03 ALFAH57987.MD
N-(3,5-Difluorophenyl)-2-(trifluoromethyl)benzamide, 97%
Purity: 97%
Molecular Formula: C14H8F5NO
Formula Weight: 301.21
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR