N-(3,5-Dimethylphenyl)-2-(trifluoromethyl)benzamide, 97%

H57451.03EA 0 EUR
ALFAH57451.03 ALFAH57451.MD
N-(3,5-Dimethylphenyl)-2-(trifluoromethyl)benzamide, 97%
Purity: 97%
Molecular Formula: C16H14F3NO
Formula Weight: 293.28
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR