3,4-Difluorobenzamidoxime ≥97%

Supplier: Thermo Scientific
Warning

H54636.03EA 90.3 EUR
H54636.03
3,4-Difluorobenzamidoxime ≥97%
3,4-Difluorobenzamidoxime
Formula: C₇H₆F₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD06660194
CAS Number: 500024-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR