N-Benzyl-3-borono-4-methylbenzenesulphonamide ≥97%
|
Warning
|
Synonyms:
5-Benzylsulphamoyl-2-methylbenzeneboronic acid
H52919.MDEA
153
EUR
H52919.MD
N-Benzyl-3-borono-4-methylbenzenesulphonamide ≥97%
N-Benzyl 3-borono-4-methylbenzenesulphonamide
Formula:
C₁₄H₁₆BNO₄S MW: 305,16 g/mol Melting Pt: 118…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD07363760 CAS Number: 871329-73-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...