2,4-Dimethoxy-4'-hydroxybenzophenone

Supplier: Molekula

MOLE13872443-5GEA 0 EUR
MOLE13872443-5G
2,4-Dimethoxy-4'-hydroxybenzophenone
2,4-Dimethoxy-4'-hydroxybenzophenone
Formula: C₁₅H₁₄O₄
MW: 258,27
MDL Number: MFCD00137850
CAS Number: 41351-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR