2-Fluoro-4'-methoxybenzophenone ≥99%

Supplier: Thermo Scientific

Warning

L13653.14EA 0 EUR
L13653.14
2-Fluoro-4'-methoxybenzophenone ≥99%
2-Fluoro-4'-methoxybenzophenone
Formula: C₁₄H₁₁FO₂
Boiling Pt: 155…160 °C (5 mmHg)
Melting Pt: 55…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00749331
CAS Number: 66938-29-2
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR