3-Benzylsulfamoyl-4-methoxybenzeneboronic acid, 93%

H53142.MDEA 95.1 EUR
ALFAH53142.MD
3-Benzylsulfamoyl-4-methoxybenzeneboronic acid, 93%
Purity: 93%
CAS No.: 874219-51-9
Molecular Formula: C14H16BNO5S
Formula Weight: 321.15
Melting Point: 134-136°
MDL No.: MFCD08235069
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR