4-(Benzylsulfamoyl)benzeneboronic acid, 95%

H53105.MDEA 41.7 EUR
ALFAH53105.MD ALFAH53105.03
4-(Benzylsulfamoyl)benzeneboronic acid, 95%
Purity: 95%
CAS No.: 548769-96-6
Molecular Formula: C13H14BNO4S
Formula Weight: 291.12
Melting Point: 148-150°
MDL No.: MFCD06659845
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR