Disulphoton sulphoxide, Supelco®

Supplier: Merck


M-Clarity™ quality level = MQ100

31562-100MG
31562-100MGEA 163 EUR
31562-100MG
Disulphoton sulphoxide, Supelco®
Disulfoton-sulphoxide
Formula: C₈H₁₉O₃PS₃
CAS Number: 2497-07-6
EINECS: 219-679-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=701e75306fddd8eb4a6540d5fb58eec55322bb5b&vid=701e75306fddd8eb4a6540d5fb58eec55322bb5b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR