2,3,3-Trimethylindolenine

Supplier: Molekula

MOLE23843892-25GEA 0 EUR
MOLE23843892-25G
2,3,3-Trimethylindolenine
2,3,3-Trimethylindolenine
Formula: C₁₁H₁₃N
MW: 159,23 g/mol
MDL Number: MFCD00005724
CAS Number: 1640-39-7
EINECS: 216-685-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR