3-Ethylbenzophenone
Supplier: LGC Standards PROMOCHEM
PROOMM0001.02EA
1600
EUR
PROOMM0001.02
3-Ethylbenzophenone
3-Ethylbenzophenone
Formula:
C₁₅H₁₄O MW: 210,2711 g/mol |
MDL Number:
MFCD01662328 CAS Number: 66067-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...