Mesitylacetic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Mesitylacetic acid

L00334.14EA 378 EUR
L00334.14 L00334.03 L00334.06
Mesitylacetic acid ≥98%
Mesitylacetic acid
Formula: C₁₁H₁₄O₂
MW: 178,23 g/mol
Melting Pt: 167…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00014368
CAS Number: 4408-60-0
EINECS: 224-556-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR