4,6-Difluoro-m-toluic acid 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2,4-Difluoro-5-methylbenzoic acid
H31770.03EA
176
EUR
H31770.03
4,6-Difluoro-m-toluic acid 98%
4,6-Difluoro-m-toluic acid
Formula:
C₈H₆F₂O₂ MW: 172,13 g/mol Melting Pt: 159…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD09835285 CAS Number: 367954-99-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...