D-(+)-Turanose

Supplier: Molekula

Synonyms: (3S,4R,5R)-1,4,5,6-Tetrahydroxy-3-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexan-2-oneTuranose

MOLEM41949862EA 0 EUR
MOLEM41949862
D-(+)-Turanose
D(+)-Turanose
Formula: C₁₂H₂₂O₁₁
MW: 342,30 g/mol
MDL Number: MFCD00006606
CAS Number: 547-25-1
EINECS: 208-918-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR