Trimethylamine-N-oxide dihydrate

Supplier: Molekula

MOLEM23966397EA 0 EUR
MOLEM23966397
Trimethylamine-N-oxide dihydrate
Trimethylamine N-oxide dihydrate
Formula: C₃H₉NO·2H₂O
MW: 111,14 (75,11 anhy)
MDL Number: MFCD00149077
CAS Number: 62637-93-8
EINECS: 214-675-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR