6-Nitro-o-anisidine ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 2-Methoxy-6-nitrophenylamine2-Methoxy-6-nitroaniline

H35777.03EA 151 EUR
H35777.03 H35777.MD
6-Nitro-o-anisidine ≥97%
6-Nitro-o-anisidine
Formula: C₇H₈N₂O₃
MW: 168,15
Melting Pt: 73…78 °C
Storage Temperature: Ambient
MDL Number: MFCD01930197
CAS Number: 16554-45-3
EINECS: 240-617-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR