2,4,6-T (2,4,6-trichlorophenoxy acetic acid)

Supplier: LGC Standards PROMOCHEM

PROONMIAP1375EA 262 EUR
PROONMIAP1375
2,4,6-T (2,4,6-trichlorophenoxy acetic acid)
2,4,6-T (2,4,6-trichlorophenoxy acetic acid)
Formula: C₈H₅Cl₃O₃
MDL Number: MFCD00053498
CAS Number: 575-89-3
EINECS: 209-394-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR