2-(2,4,5-Trichlorophenoxy)propionic acid 96%

Supplier: Thermo Scientific
Warning

L08136.14EA 229 EUR
L08136.14 L08136.06
2-(2,4,5-Trichlorophenoxy)propionic acid 96%
2-(2,4,5-Trichlorophenoxy)propionic acid
Formula: C₉H₇Cl₃O₃
MW: 269,51
Melting Pt: 176…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00002646
CAS Number: 93-72-1
EINECS: 202-271-2
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR