2-Methyl-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: Thermo Scientific
Warning

H26628.06EA 487 EUR
H26628.06 H26628.03
2-Methyl-5-(trifluoromethyl)benzaldehyde ≥97%
2-Methyl-5-(trifluoromethyl)benzaldehyde
Formula: C₉H₇F₃O
Storage Temperature: Ambient
MDL Number: MFCD06660200
CAS Number: 886498-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR