L(-)-Fucose

Supplier: Molekula

Synonyms: 6-Deoxy-L-Galactose

MOLEM21002013EA 0 EUR
MOLEM21002013
L(-)-Fucose
L(-)-Fucose
Formula: C₆H₁₂O₅
MW: 164,16 g/mol
MDL Number: MFCD00069812
CAS Number: 2438-80-4
EINECS: 219-452-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR