Iron(III) chloride hexahydrate, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Iron trichloride hexahydrateFerric chloride hexahydrate

0175.0100EA 46.93 EUR
0175.0100 0175.1000
Iron(III) chloride hexahydrate, BAKER ANALYZED® ACS, J.T.Baker®
Iron (III) chloride hexahydrate
Formula: FeCl₃·6H₂O
MW: 270,3 g/mol
Boiling Pt: 280…285 °C (1013 hPa)
Melting Pt: 37 °C
Density: 1,82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149712
CAS Number: 10025-77-1
EINECS: 231-729-4
UN: 3260
ADR: 8,III
Merck Index: 13,04048
https://ie.vwr.com/stibo/web/std.lang.all/05/51/16640551.jpg

Order Now

Specification Test Results

Meets ACS Specifications
Assay (by Iodometry) 97.0 - 102.0 %
Copper (Cu) (by AAS) max. 0.003 %
Calcium (Ca) max. 0.01 %
Potassium (K) max. 0.005 %
Magnesium (Mg) max. 0.005 %
Sodium (Na) max. 0.05 %
Ferrous Iron (as Fe) max. 0.002 %
Insoluble Matter max. 0.01 %
Nitrate (NO3) max. 0.01 %
Phosphorus Compounds (as PO4) max. 0.01 %
Sulfate (SO4) max. 0.01 %
Zinc (Zn) (by AAS) max. 0.003 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR