4'-Isobutylacetophenone
Supplier: LGC Standards PROMOCHEM
Synonyms:
p-Isobutylacetophenone
PROOMM0002.04EA
1220
EUR
PROOMM0002.04
4'-Isobutylacetophenone
4'-Isobutylacetophenone
Formula:
C₁₂H₁₆O |
MDL Number:
MFCD00027393 CAS Number: 38861-78-8 EINECS: 254-159-8 UN: 1224 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...