2-Methoxyestradiol

Supplier: Molekula

MOLE25868681-25MGEA 0 EUR
MOLE25868681-25MG
2-Methoxyestradiol
2-Methoxyestradiol
Formula: C₁₉H₂₆O₃
MDL Number: MFCD00010489
CAS Number: 362-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR