Arachidonic acid
Supplier: Cayman Chemical
Synonyms:
(5Z,8Z,11Z,14Z)-Icosa-5,8,11,14-tetraenoic acid
CAYM10010377-1EA
197
EUR
CAYM10010377-1
Arachidonic acid
Arachidonic acid
Formula:
C₂₀H₃₂O₂ MW: 304,5 g/mol |
MDL Number:
MFCD00004417 CAS Number: 506-32-1 EINECS: 208-033-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...