α-D(+)-Talose 97%

Supplier: Thermo Scientific

B21226.02 B21226.77
B21226.02EA 450 EUR
B21226.02 B21226.77
α-D(+)-Talose 97%
α-D-Talose
Formula: C₆H₁₂O₆
MW: 180,16 g/mol
Melting Pt: 133…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00064906
CAS Number: 2595-98-4
EINECS: 219-996-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR