D-(-)-Lyxose

Supplier: Molekula

MOLE10556383-5GEA 0 EUR
MOLE10556383-5G
D-(-)-Lyxose
D(-)-Lyxose
Formula: C₅H₁₀O₅
MW: 150,13
Storage Temperature: Ambient
MDL Number: MFCD00064362
CAS Number: 1114-34-7
EINECS: 214-212-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR